Skip to Content
Merck
All Photos(1)

Documents

Safety Information

P5539

Sigma-Aldrich

Anti-Prostaglandin F antibody produced in rabbit

whole antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

storage temp.

2-8°C

Immunogen

PGF−BSA

Reconstitution

Reconstitute with 5 mL of 0.01 M phosphate buffered saline, pH 7.4, containing 0.1% bovine serum albumin and 0.1% sodium azide.

Analysis Note

Tests per vial and specificity are determined using a dextran coated charcoal radioimmunoassay (RIA).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

P5539-VAR:
P5539-10X100TST:
P5539-100X100TST:
P5539-100TST:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Siemieniuch et al.
Polish journal of veterinary sciences, 13(4), 605-613 (2010-01-01)
Tumor Necrosis Factor-alpha (TNFalpha) is a pleiotrophic cytokine, affects either normal or tumor cells, and influences cellular differentiation. TNFalpha role in female reproduction has been proven to be mediated through an influence on prostaglandin (PGs) synthesis and output. To evaluate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service