Skip to Content
Merck
All Photos(1)

Key Documents

P4118

Sigma-Aldrich

[CPP1-7, NPY19-23, Ala31, Aib32, Gln34]-Pancreatic polypeptide

Synonym(s):

GPSQPTYPGDNATPEQMARYYSALRRYINMA-Aib-RQRY

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C183H281N57O54S2
Molecular Weight:
4207.67
MDL number:
UNSPSC Code:
12352202

form

solid

composition

Peptide content, ≥80%

storage condition

desiccated

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NPY(4852)

Amino Acid Sequence

Gly-Pro-Ser-Gln-Pro-Thr-Tyr-Pro-Gly-Asp-Asn-Ala-Thr-Pro-Glu-Gln-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-Arg-Tyr-Ile-Asn-Met-Ala-Aib-Arg-Gln-Arg-Tyr

Biochem/physiol Actions

First potent and selective Y5 neuropeptide Y receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service