Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

O5505

Sigma-Aldrich

Stearyl oleate

~99%

Synonym(s):

Oleic acid stearyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H70O2
CAS Number:
Molecular Weight:
534.94
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C36H70O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-38-36(37)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h18,20H,3-17,19,21-35H2,1-2H3/b20-18-

InChI key

WRPMUZXHQKAAIC-ZZEZOPTASA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

O5505-VAR:
O5505-1G:
O5505-100MG:
O5505-500MG:
O5505-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E Stewart et al.
Lipids, 16(5), 355-359 (1981-05-01)
Chromatography of stearyl oleate and cholesteryl oleate on thin layer plates coated with magnesium hydroxide-celite, 1:1, or magnesia-celite, 1:1, showed that magnesium hydroxide had better resolving power for the separation of these wax ester and steryl ester model compounds than
Jianzhong Chen et al.
Lipids, 51(11), 1269-1287 (2016-10-25)
Wax esters (WE) are one of the predominant lipid types in human meibomian gland secretions (meibum) and account for 40-50 % of total meibum lipids. Recently, we managed to quantify 51 isomeric groups of intact WE in normal human meibum samples
Andrey B Imbs et al.
PloS one, 14(4), e0215759-e0215759 (2019-04-30)
To expand our knowledge of lipid and fatty acid (FA) biosynthesis in marine cnidarians, polar lipidomes of hydrocorals were studied for the first time and then compared with those of soft corals from tropical and boreal regions. The structure and
Jianzhong Chen et al.
Lipids, 50(8), 821-836 (2015-07-17)
A series of different types of wax esters (represented by RCOOR') were systematically studied by using electrospray ionization (ESI) collision-induced dissociation tandem mass spectrometry (MS/MS) along with pseudo MS(3) (in-source dissociation combined with MS/MS) on a quadrupole time-of-flight (Q-TOF) mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service