Skip to Content
Merck
All Photos(1)

Key Documents

O3632

Sigma-Aldrich

Osteocalcin Fragment 7-19 human

≥97% (HPLC)

Synonym(s):

Gly-Ala-Pro-Val-Pro-Tyr-Pro-Asp-Pro-Leu-Glu-Pro-Arg

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H98N16O19
CAS Number:
Molecular Weight:
1407.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C1CCCN1C(=O)C(CC(O)=O)NC(=O)C2CCCN2C(=O)C(Cc3ccc(O)cc3)NC(=O)C4CCCN4C(=O)C(NC(=O)C5CCCN5C(=O)C(C)NC(=O)CN)C(C)C)C(=O)NC(CCC(O)=O)C(=O)N6CCCC6C(=O)NC(CCCNC(N)=N)C(O)=O

InChI

1S/C65H98N16O19/c1-34(2)30-41(53(88)71-39(22-23-50(84)85)60(95)78-26-8-13-44(78)54(89)72-40(64(99)100)12-6-24-69-65(67)68)73-55(90)45-14-9-28-80(45)62(97)43(32-51(86)87)75-56(91)46-15-10-27-79(46)61(96)42(31-37-18-20-38(82)21-19-37)74-57(92)47-16-11-29-81(47)63(98)52(35(3)4)76-58(93)48-17-7-25-77(48)59(94)36(5)70-49(83)33-66/h18-21,34-36,39-48,52,82H,6-17,22-33,66H2,1-5H3,(H,70,83)(H,71,88)(H,72,89)(H,73,90)(H,74,92)(H,75,91)(H,76,93)(H,84,85)(H,86,87)(H,99,100)(H4,67,68,69)

InChI key

XEVUUFOXAJNOGZ-UHFFFAOYSA-N

Gene Information

human ... BGLAP(632)

Looking for similar products? Visit Product Comparison Guide

Other Notes

Calcium-binding protein fragment.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P V Hauschka
Haemostasis, 16(3-4), 258-272 (1986-01-01)
Osteocalcin is an abundant Ca2+-binding protein indigenous to the organic matrix of bone, dentin, and possibly other mineralized tissues. This protein contains 47-50 amino acid residues (molecular weight 5,200-5,900) depending on the species. Osteocalcin is distinguished by its content of
H Jüppner et al.
Calcified tissue international, 39(5), 310-315 (1986-11-01)
Radioiodination of synthetic human 37-49 osteocalcin requires optimal labeling conditions in order to obtain a maximum of mono- and di-iodinated tracer with little contamination by tri- and tetra-iodinated products or "radio-damage." The antibody raised against osteocalcin(h37-49) had the highest affinity
P Pastoureau et al.
Clinical chemistry, 36(9), 1620-1624 (1990-09-01)
Most RIAs of serum bone gla-protein (BGP; also called osteocalcin) used for clinical investigation are based on bovine BGP for standard, tracer, and immunogen because of the homology between bovine and human BGP. However, ovine BGP differs from human BGP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service