Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

N8136

Sigma-Aldrich

Neocarratetraose 41,43-disulfate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C24H36O25S2Na2
CAS Number:
Molecular Weight:
834.64
UNSPSC Code:
12352201
PubChem Substance ID:

form

solid

storage temp.

2-8°C

SMILES string

[Na+].[Na+].OC[C@H]1O[C@@H](O)[C@H](O)[C@@H](O[C@H]2O[C@@H]3CO[C@H]([C@H]2O)C3O[C@@H]4O[C@H](CO)[C@H](OS([O-])(=O)=O)[C@H](O[C@H]5O[C@@H]6CO[C@@H](C6O)[C@H]5O)[C@H]4O)[C@H]1OS([O-])(=O)=O

InChI

1S/C24H38O25S2.2Na/c25-1-5-15(48-50(33,34)35)19(10(28)21(32)41-5)46-23-12(30)18-14(8(44-23)4-40-18)45-24-13(31)20(16(6(2-26)42-24)49-51(36,37)38)47-22-11(29)17-9(27)7(43-22)3-39-17;;/h5-32H,1-4H2,(H,33,34,35)(H,36,37,38);;/q;2*+1/p-2/t5-,6-,7-,8-,9?,10-,11-,12-,13-,14?,15+,16+,17+,18-,19-,20-,21-,22-,23-,24+;;/m1../s1

InChI key

VJDGBWQZCPEBMZ-PHERPFAOSA-L

Other Notes

Mixed anomers

Preparation Note

Prepared enzymatically from κ-carrageenan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

N8136-BULK:
N8136-5MG:
N8136-100MG:
N8136-25MG:
N8136-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M W McLean et al.
European journal of biochemistry, 93(3), 553-558 (1979-02-01)
A kappa-carrageenase was isolated from the cell-free medium of cultured Pseudomonas carrageenovora. From dodecylsulphate/polyacrylamide gel electrophoresis, a single protein (identified as the kappa-carrageenase) was detected in the medium. Activity against nominal carrageenan types and inspection of the products indicate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service