Skip to Content
Merck
All Photos(1)

Documents

Safety Information

MAK496

Sigma-Aldrich

Zinc Assay Kit

sufficient for 250 colorimetric tests

Synonym(s):

Zinc Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

environmental
pharmaceutical

detection method

colorimetric

relevant disease(s)

diabetes; renal disease; immunological diseases; gastrointestinal diseases; cancer; hematological disorder; genitourinary disorders

shipped in

wet ice

storage temp.

−20°C

Related Categories

General description

Zinc is an essential trace element and plays many key roles in metabolism. It is required for the activity of more than 300 enzymes, the structure of many proteins, and control of genetic expression. Zinc status affects basic processes of cell division, growth, differentiation, development, performance and aging through its requirement for synthesis and repair of DNA, RNA and proteins. The common causes of zinc deficiency are low dietary intakes and low bioavailability. Clinical signs of zinc deficiency include acrodermatitis, low immunity, diarrhea, poor healing, stunting, hypogonadism, fetal growth failure, teratology and abortion. Zinc deficiency has now been recognized to be associated with many diseases such as malabsorption syndrome, chronic liver disease, chronic renal disease, sickle cell disease, diabetes, malignancy, and other chronic illnesses.

Features and Benefits

  • Linear detection range 0.12 µM (0.78 µg/dL) to 10 µM (65 µg/dL) zinc
  • Compatible with high-throughput handling systems.
  • The optimized formulation has greatly enhanced reagent and signal stability.

Suitability

The kit is suitable for the determination of zinc in serum, plasma, urine, saliva, waste water, soil, etc, as well as for studying the effects of drugs on zinc metabolism.

Principle

The Zinc Assay Kit is designed to measure zinc directly in biological samples without any pretreatment. The method utilizes a chromogen that forms a colored complex specifically with zinc. The intensity of the color, measured at 425 nm, is directly proportional to the zinc concentration in the sample.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Please refer to KIT Component information

PRTR

Please refer to KIT Component information

FSL

Please refer to KIT Component information

ISHL Indicated Name

Please refer to KIT Component information

ISHL Notified Names

Please refer to KIT Component information

Cartagena Act

Please refer to KIT Component information

JAN Code

キットコンポーネントの情報を参照してください


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service