MAK474
Acetate Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
Acetate Detection Kit
About This Item
Recommended Products
Quality Level
application(s)
agriculture
environmental
food and beverages
pharmaceutical
detection method
colorimetric
fluorometric
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Features and Benefits
- Detection range: 0.20 to 20 mM for the colorimetric assay and 0.13 to 2 mM for the fluorometric assay.
Suitability
Principle
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Please refer to KIT Component information
PRTR
Please refer to KIT Component information
FSL
Please refer to KIT Component information
ISHL Indicated Name
Please refer to KIT Component information
ISHL Notified Names
Please refer to KIT Component information
Cartagena Act
Please refer to KIT Component information
JAN Code
キットコンポーネントの情報を参照してください
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service