Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

M5685

Sigma-Aldrich

Metrazoline

solid

Synonym(s):

4,5-Dihydro-2-[(1E)-2-(2-methylphenyl)ethenyl-1H-imidazole oxalate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H14N2·C2H2O4
CAS Number:
Molecular Weight:
276.29
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

solubility

H2O: 16 mg/mL

SMILES string

CC1=CC=CC=C1/C=C/C2=NCCN2.OC(C(O)=O)=O

InChI

1S/C12H14N2.C2H2O4/c1-10-4-2-3-5-11(10)6-7-12-13-8-9-14-12;3-1(4)2(5)6/h2-7H,8-9H2,1H3,(H,13,14);(H,3,4)(H,5,6)/b7-6+;

InChI key

QDUUQIGHAAEJDO-UHDJGPCESA-N

Biochem/physiol Actions

Potent and selective imidazoline binding site ligand

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

M5685-25MG:4548173202990
M5685-BULK:
M5685-VAR:
M5685-5MG:4548173203003


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Polidori et al.
European journal of pharmacology, 392(1-2), 41-49 (2000-04-05)
Previous studies have suggested that imidazoline I(2) receptors play a role in feeding control in rats. The effect of subcutaneous (s.c.) injections of four novel imidazoline I(2) ligands, 2-naphthalen-2yl-4,5-dihydro-1H-imidazole hydrochloride (benazoline), 2-styryl-4,5-dihydro-1H-imidazole oxalate (tracizoline), o-nitro-tracizoline and o-methyl-tracizoline (metrazoline) on food

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service