IRC50
Amberlite™ Weakly Acidic Cation Exchanger, Hydrogen form
macroreticular
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
47101511
Recommended Products
form
macroreticular
contains
~48% moisture
matrix
4% cross-linked polystyrene
matrix active group
carboxylic acid
particle size
16-50 mesh (wet)
capacity
10.0 meq/g
3.5 meq/mL
Looking for similar products? Visit Product Comparison Guide
Legal Information
Amberlite is a trademark of DuPont de Nemours, Inc.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service