Key Documents
G2775
Sigma-Aldrich
Gaskets for vertical mini gel
10 cm wide, size 1mm x 8cm, color-coded red
About This Item
UNSPSC Code:
12352200
NACRES:
NB.51
Product Name
Gaskets for vertical mini gel 10 cm wide, size 1 mm × 8 cm, color-coded red
material
silicone
Quality Level
size
1 mm × 8 cm
color
color-coded red
Features and Benefits
Unique gasket provides continuous seal around sides and bottom of glass plates, eliminating the need for a bottom spacer. Gasket thickness must match comb and spacers. Length of gasket material is approx. 26 cm (2 × h + w). Gaskets are color-coded for easy identification.
Choose from one of the most recent versions:
Certificates of Analysis (COA)