Skip to Content
Merck
All Photos(1)

Documents

F6128

Sigma-Aldrich

N-Formyl-Met-Trp

≥90% (HPLC), crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3O4S
CAS Number:
Molecular Weight:
363.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥90% (HPLC)

form

crystalline

color

off-white to tan

storage temp.

−20°C

SMILES string

CSCCC(NC=O)C(=O)NC(Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C17H21N3O4S/c1-25-7-6-14(19-10-21)16(22)20-15(17(23)24)8-11-9-18-13-5-3-2-4-12(11)13/h2-5,9-10,14-15,18H,6-8H2,1H3,(H,19,21)(H,20,22)(H,23,24)

InChI key

AAIWVNZNCMTTQF-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A B Edmundson et al.
Molecular immunology, 22(4), 463-475 (1985-04-01)
X-ray crystallographic techniques were used to study the modes of binding of N-formylated chemotactic peptides to the Mcg light chain (Bence-Jones) dimer. By difference Fourier analyses at 2.7-A resolution four N-formylated tripeptides were found to occupy similar sites in the
S K Ainsworth et al.
The American review of respiratory disease, 124(3), 280-284 (1981-09-01)
Through the application of a simplified in vitro assay for chemotaxis of polymorphonuclear leukocytes, potent chemotaxins have been found to exist in cotton mill dust extracts. These substances may be involved in the recruitment of cells to pulmonary surfaces and
U Moser et al.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition, 54(1), 47-53 (1984-01-01)
The uptake of ascorbic acid by isolated human granulocytes is investigated under different conditions. The rate of uptake depends on the concentration of ascorbic acid in the incubation medium as well as on temperature, with a maximum at 40 degrees

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service