EP1101
Sigma-Aldrich® MSMINIDUO horizontal gel electrophoresis system
capable of resolving up to 48 different samples
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
grade
electrophoresis grade
usage
up to 32 samples for 7x7 cm tray
up to 64 samples for 7x10 cm tray
W × L × H
21 cm × 9 cm × 9 cm, unit dimensions
volume
225 mL , buffer volume
Application
This is the smallest unit in the range, designed for the researcher with low to medium numbers of samples. Unit includes 7 x 7 cm and 7 x 10 cm UV Tray, 2 x 8 sample, 1 mm thick combs, casting dams, and loading guides. The small gel size maximizes run economy but does not compromise versatility as two tray options are included. The combs range from preparative up to 16 samples. Simply by altering the gel tray or comb, this compact unit is capable of resolving up to 48 different samples, prepping 1 mL of sample or separating sample bands over a distance of 9 cm.
Components
Corrosion resistant gold plated lid terminals with retractable connectors compatible with most major power supplies.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service