Skip to Content
Merck
All Photos(1)

Key Documents

EHU033811

Sigma-Aldrich

MISSION® esiRNA

targeting human P2RY1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
¥38,100
50 μG
¥68,300

¥38,100


Check Cart for Availability


Select a Size

Change View
20 μG
¥38,100
50 μG
¥68,300

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

¥38,100


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCATCTGGATGTTCGTCTTCCACATGAAGCCCTGGAGCGGCATCTCCGTGTACATGTTCAATTTGGCTCTGGCCGACTTCTTGTACGTGCTGACTCTGCCAGCCCTGATCTTCTACTACTTCAATAAAACAGACTGGATCTTCGGGGATGCCATGTGTAAACTGCAGAGGTTCATCTTTCATGTGAACCTCTATGGCAGCATCTTGTTTCTGACATGCATCAGTGCCCACCGGTACAGCGGTGTGGTGTACCCCCTCAAGTCCCTGGGCCGGCTCAAAAAGAAGAATGCGATCTGTATCAGCGTGCTGGTGTGGCTCATTGTGGTGGTGGCGATCTCCCCCATCCTCTTCTACTCAGGTACCGGGGTCCGCAAAAACAAAACCATCACCTGTTACGACACCACCTCAGACG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Audrey Castaing-Berthou et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 42(2), 579-593 (2017-06-05)
High-density lipoproteins (HDL) exert multiple cardioprotective functions on the arterial wall, including the promotion of endothelial cell survival and proliferation. Among mechanism contributing to endothelial protection, it has been reported that apolipoprotein A-I (apoA-I), the major protein in HDL, binds
Maosheng Xia et al.
Journal of neuroscience research, 92(8), 1078-1090 (2014-04-02)
After spinal cord injury (SCI), the formation of glial scar is a complex process that is attributed primarily to astrocytic proliferation, but the mechanism of astrocytes proliferation is still unclear. Fibronectin is a large extracellular glycoprotein that helps organize the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service