Skip to Content
Merck
All Photos(1)

Key Documents

D2005

Sigma-Aldrich

DNP-L-tryptophan

Synonym(s):

N-(2,4-Dinitrophenyl)-L-tryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N4O6
CAS Number:
Molecular Weight:
370.32
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

OC(=O)C(Cc1c[nH]c2ccccc12)Nc3ccc(cc3N(=O)=O)N(=O)=O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service