Skip to Content
Merck
All Photos(4)

Key Documents

Safety Information

CWA-1139

Sigma-Aldrich

Anti-Mouse Integrin α5/CD49e (5H10-27) ColorWheel® Dye-Ready mAb

greener alternative

Synonym(s):

Integrin alpha-5;CD49 antigen-like family member E;Fibronectin receptor subunit alpha;Integrin alpha-F;VLA-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5H10-27, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 115.04 kDa

purified by

affinity chromatography

species reactivity

mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Itga5(16402)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone 5H10-27 is a rat monoclonal antibody that detects mouse Integrin 5 (CD49e). It targets an epitope within the extracellular domain.

Immunogen

C57BL/6 x A/J F1 mouse mast cell line MC/9

Application

Quality Control Testing

Evaluated by Flow Cytometry in C57Bl/6 bone marrow cells .

Staining of one million C57BL/6 mouse bone marrow cells was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1139, Anti-Mouse Integrin 5/CD49e (5H10-27) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or a 1:1 mixture of an equivalent amount of Cat. No. CWA-1018 Rat IgG2a isotype control and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye.

Tested Application

Flow Cytometry Analysis: Staining of one million C57BL/6 mouse bone marrow cells was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1139, Anti-Mouse Integrin 5/CD49e (5H10-27) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1018 Rat IgG2a isotype control and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Integrin alpha-5 (UniProt: P11688; also known as CD49 antigen-like family member E, Fibronectin receptor subunit alpha, Integrin alpha-F, VLA-5, CD49e) is encoded by the Itga5 gene (Gene ID: 16402) in murine species. Integrins are heterodimeric integral membrane proteins composed of an -subunit and a β-subunit that function in cell surface adhesion and signaling. Integrin 5/CD49e is expressed on thymocytes, T cells, monocytes, platelets, early B cells, and activated in B cells. It exhibits an increased expression in mesodermal progenitors and is shown to be prevalent in fetal cardiomyocytes. Its deletion is shown to cause mesodermal defects during embryogenesis in murine models. Membrane fractions obtained from fibroblasts of diabetic animals display significant reduction in integrin 5/CD49e levels. Integrin 5/CD49e is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-44), which is subsequently cleaved off to generate the mature form that contains a heavy chain (aa 45-898) and a light chain (aa 899-1053). Structurally, it consists of an extracellular domain (aa 45-999), a transmembrane domain (aa 1000-1025), and a short cytoplasmic domain (aa 1026-1053). It contains four calcium binding sites (aa 480-488; 334-342; 401-409; and 465-473). When associated with β1 subunit it can serve as a receptor for fibronectin and fibrinogen. It recognizes the sequence R-G-D in its ligands. Integrin 5/β1 is reported to bind to PLA2G2A via a site (site 2) that is distinct from the classical ligand-binding site (site 1) and this induces integrin conformational changes and enhanced ligand binding to site 1. (Ref.: Neiman, G., et al. (2019). Sci. Rep. 9; 18077; Su, Y., et al. (2016). Proc. Natl. Acad. Sci. USA. 113(27); E3872-E3881).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

CWA-1139-25UL:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service