Skip to Content
Merck
All Photos(1)

Documents

CLS355467

Falcon® Polystyrene Plate

well size 6 wells

Synonym(s):

Cell Culture Plate, plastic coated cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13

feature

lid: no

packaging

pkg of 4 ea (1 pack of 4)

manufacturer/tradename

Corning 355467

well size

6 wells

binding type

Tissue Culture (TC)-treated surface

Application

Cell culture

Legal Information

Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service