Skip to Content
Merck
All Photos(1)

Documents

NCTC 4206

91010303, hamster peritoneum, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCTC 4206, 91010303

biological source

hamster peritoneum

growth mode

Adherent

karyotype

2n = 22; Modal no. 23

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese peritoneum

Cell Line Description

A chemically defined medium derivative of B14FAF28-G3. Since it was established, NCTC cells have been employed in nutritional studies involving 6 different NCTC chemically defined serum-free media.

Application

Nutritional studies

Culture Medium

NCTC 135 100% + 2mM Glutamine.

Subculture Routine

Sub-cultures are prepared by scraping; 5% CO2; 37°C. Media change three times a week. Freeze in 9% glycerol/91% FBS.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service