Skip to Content
Merck
All Photos(1)

Documents

SJK-287-38

90112228, Reactivity: Anti-DNA Polymerase, human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

(BALB/c x C57BL/6)F1 mouse spleen

Myeloma

NS1

Reactivity

Anti-DNA Polymerase, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse NS1 x Mouse (BALB/c x C57BL/6)F1 spleen

Cell Line Description

The antibody binds KB cell DNA polymerase and exhibits potent neutralising activity against DNA polymerase _.

Immunogen

KB cell DNA polymerase fraction VIII

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service