Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

C2242

Supelco

Chemichrome Western Control

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

description

Colored Molecular Weight Markers

storage temp.

−20°C

Application

Chemichrome™ Western Control is an ideal molecular weight marker and positive control for researchers performing electrophoresis and subsequent Western blotting. Similar to Colorburst™, Chemichrome contains an additional band of mouse IgG. During electrophoresis, brightly colored protein bands serve as positive controls for protein migration. During blotting, the same brightly colored bands indicate that the transfer is complete. After incubation with mouse primary and secondary antibodies, the band of mouse IgG serves as a positive control using either colorimetric or chemiluminescent substrates. Chemichrome is supplied in vials containing 200 μl of ready-to-use solution.

Features and Benefits

  • Confirms that a membrane transfer is complete
  • Confirms successful Western Blotting Conditions
  • Compatible with many peroxidase and phosphatase substrates, including TMB and ECL®.
·

Legal Information

Chemichrome is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances
Class II Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

C2242-BULK:
C2242PROC:
C2242-VAR:
C2242-1VL:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service