C0894
Cinchonidine hydrochloride
Synonym(s):
Cinchonan-9(R)-ol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H22N2O · HCl
CAS Number:
Molecular Weight:
330.85
EC Number:
MDL number:
UNSPSC Code:
41116107
Recommended Products
SMILES string
Cl.O[C@@H]([C@@H]1C[C@@H]2CC[N@H]1C[C@@H]2C=C)c3ccnc4ccccc34
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service