Documents
BMSIPD960015PTFB
Sigma-Aldrich
BeadBlaster 96 PTFE Grinding ball with Steel core
bead diam. 15 mm, BeadBlaster 96
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
material
PTFE (with steel core)
packaging
pack of 2
manufacturer/tradename
Benchmark Scientific IPD9600-15PTFB
bead diam.
15 mm
compatibility
BeadBlaster 96
Related Categories
General description
PTFE is the least hard of the grinding media materials and is chosen for its inert properties. PTFE is metal-free and non-stick. It′s high temperature resistance expands it′s use to cryogrinding. PTFE grinding media is used for applications in the pharmaceutical field as well as for plants and other biological samples. The 15mm PTFE grinding balls have steel cores for a higher energy input.
Application
Homogenization of biological and pharmaceutical samples
Features and Benefits
  • Inert material
  • Non-stick
  • Wide temperature stability
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.