Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR96LC480WBC

PCR microplate for Roche® 480 LightCycler®

barcoded, with UC-500 film, size 96 wells, pkg of 5x10plates/cs, white

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
96 wells
Product AXYPCR96LC480WBC is not currently sold in your country. Contact Technical Service

material

conical bottom wells

manufacturer/tradename

Axygen Inc PCR-96-LC480-W-BC
Corning

packaging

pkg of 5x10plates/cs

size

96 wells

color

white

Looking for similar products? Visit Product Comparison Guide

Legal Information

LightCycler is a registered trademark of Roche
Roche is a registered trademark of Roche Molecular Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service