Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

AV53826

Sigma-Aldrich

Anti-LPIN1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp781P1796, Anti-KIAA0188, Anti-Lipin 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
¥62,700

¥62,700


Estimated to ship onMay 04, 2025

New, lower price on this item!


Select a Size

Change View
100 μL
¥62,700

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

¥62,700


Estimated to ship onMay 04, 2025

New, lower price on this item!

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

98 kDa

species reactivity

horse, human, mouse, rabbit, dog, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LPIN1(23175)

Immunogen

Synthetic peptide directed towards the N terminal region of human LPIN1

Application

Anti-LPIN1 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

LPIN1 (lipin 1) encodes a magnesium-ion-dependent phosphatidic acid phosphohydrolase enzyme that catalyzes the dephosphorylation of phosphatidic acid to form diacylglycerol. It is a transcriptional co-regulator that regulates lipid metabolism and adipogenesis (adipocyte maturation and maintenance) by modulating the C/EBPα (CCAAT/enhancer-binding protein α) and PPAR γ (peroxisome-proliferator-activated receptor γ) network. Lipin 1 also plays a pivotal role in controlling autophagy clearance by facilitating the maturation of autolysosomes via stimulation of protein kinase D (PKD)-Vps34 phosphatidylinositol 3-kinase signaling cascade.

Sequence

Synthetic peptide located within the following region: SLAVIYPQSASYPNSDREWSPTPSPSGSRPSTPKSDSELVSKSTERTGQK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

AV53826-50UG:
AV53826-100UL:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peixiang Zhang et al.
Cell metabolism, 20(2), 267-279 (2014-06-17)
LPIN1 encodes lipin-1, a phosphatidic acid phosphatase (PAP) enzyme that catalyzes the dephosphorylation of phosphatidic acid to form diacylglycerol. Homozygous LPIN1 gene mutations cause severe rhabdomyolysis, and heterozygous LPIN1 missense mutations may promote statin-induced myopathy. We demonstrate that lipin-1-related myopathy
Hee Eun Kim et al.
The Biochemical journal, 453(1), 49-60 (2013-05-01)
PPARγ (peroxisome-proliferator-activated receptor γ) is a master transcription factor involved in adipogenesis through regulating adipocyte-specific gene expression. Recently, lipin1 was found to act as a key factor for adipocyte maturation and maintenance by modulating the C/EBPα (CCAAT/enhancer-binding protein α) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service