Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

A9673

Sigma-Aldrich

Arachidonic acid

from Porcine liver, ≥99% (GC), liquid

Synonym(s):

cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid, Eicosa-5Z,8Z,11Z,14Z-tetraenoic acid, Immunocytophyte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4(CH=CHCH2)4CH2CH2CO2H
CAS Number:
Molecular Weight:
304.47
Beilstein:
1713889
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

Porcine liver

Assay

≥99% (GC)

form

liquid

color

clear colorless to very faintly yellow

refractive index

n20/D 1.4872 (lit.)

bp

169-171 °C/0.15 mmHg (lit.)

mp

−49 °C (lit.)

solubility

ethanol: ≥10 mg/mL (lit.)(lit.)

density

0.922 g/mL at 25 °C (lit.)

application(s)

food and beverages

functional group

carboxylic acid

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O

InChI

1S/C20H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-,16-15-

InChI key

YZXBAPSDXZZRGB-DOFZRALJSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Arachidonic acid (AA) is an unsaturated ω6 fatty acid constituent of the phospholipids of cell membranes. Phospholipase A2 releases AA from the membrane phospholipids in response to inflammation. AA is subsequently metabolized to prostaglandins and thromboxanes by at least two cyclooxygenase (COX) isoforms, to leukotrienes and lipoxins by lipoxygenases, and to epoxyeicosatrienoic acids via cytochrome p450-catalyzed metabolism. AA and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation, and apoptosis. AA has been demonstrated to bind to the a subunit of G protein and inhibit the activity of Ras GTPase-activating proteins (GAPs). Cellular uptake of AA is energy dependent and involves protein-facilitated transport across the plasma membrane.
Arachidonic acid stimulates adhesion of MDA-MB-435 human metastatic cancer cells to extracellular matrix molecules (collagen IV and vitronectin) .

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

A9673-500MG:
A9673-1G:
A9673-BULK:
A9673-100MG:
A9673-10MG:
A9673-VAR:
A9673-50MG:
A9673-2G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingying Chen et al.
Scientific reports, 6, 26608-26608 (2016-05-21)
Definitive haematopoiesis occurs during the lifetime of an individual, which continuously replenishes all blood and immune cells. During embryonic development, haematopoietic stem cell (HSC) formation is tightly controlled by growth factors, signalling molecules and transcription factors. But little is known
Robert Heler et al.
Channels (Austin, Tex.), 7(2), 74-84 (2013-01-22)
Polyunsaturated fatty acids such as arachidonic acid (AA) exhibit inhibitory modulation of Kv4 potassium channels. Molecular docking approaches using a Kv4.2 homology model predicted a membrane-embedded binding pocket for AA comprised of the S4-S5 linker on one subunit and several
Nadia García-Mateo et al.
Frontiers in cellular neuroscience, 8, 374-374 (2014-11-27)
Management of lipids, particularly signaling lipids that control neuroinflammation, is crucial for the regeneration capability of a damaged nervous system. Knowledge of pro- and anti-inflammatory signals after nervous system injury is extensive, most of them being proteins acting through well-known
William J Gault et al.
The Journal of cell biology, 207(6), 767-782 (2014-12-24)
Osmotic cues from the environment mediate rapid detection of epithelial breaches by leukocytes in larval zebrafish tail fins. Using intravital luminescence and fluorescence microscopy, we now show that osmolarity differences between the interstitial fluid and the external environment trigger ATP
Martin Kongsbak et al.
BMC immunology, 15, 35-35 (2014-09-19)
In vitro studies have shown that the active form of vitamin D3, 1α,25-dihydroxyvitamin D3 (1,25(OH)2D3), can regulate differentiation of CD4+ T cells by inhibiting Th1 and Th17 cell differentiation and promoting Th2 and Treg cell differentiation. However, the serum concentration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service