Skip to Content
Merck
All Photos(1)

Key Documents

A9646

Sigma-Aldrich

N-Acetylneuraminic acid

from sheep submaxillary glands, ≥99%

Synonym(s):

5-Acetamido-3,5-dideoxy-D-glycero-D-galactononulosonic acid, Lactaminic acid, NAN, NANA, Sialic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO9
CAS Number:
Molecular Weight:
309.27
Beilstein:
1716283
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

sheep submaxillary glands

type

Type VIII

Assay

≥99%

storage temp.

−20°C

SMILES string

O[C@@]1(O[C@@]([C@@H]([C@H](C1)O)NC(C)=O)([H])[C@@H]([C@@H](CO)O)O)C(O)=O

InChI

1S/C11H19NO9/c1-4(14)12-7-5(15)2-11(20,10(18)19)21-9(7)8(17)6(16)3-13/h5-9,13,15-17,20H,2-3H2,1H3,(H,12,14)(H,18,19)/t5-,6+,7+,8+,9+,11-/m0/s1

InChI key

SQVRNKJHWKZAKO-PFQGKNLYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Both sialic acid and neuraminic acid are loosely used to refer to conjugates of neuraminic acid. N-Acetylneuraminic acid is often found as the terminal sugar of cell surface glycoproteins. Cell surface glycoproteins have important roles in cell recognition and interaction as well as in cell adhesion. Membrane glycoproteins are also important in tumor growth and metastases.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reshmi Parameswaran et al.
The Journal of experimental medicine, 210(4), 805-819 (2013-03-13)
The development of resistance to chemotherapy is a major cause of relapse in acute lymphoblastic leukemia (ALL). Though several mechanisms associated with drug resistance have been studied in detail, the role of carbohydrate modification remains unexplored. Here, we investigated the
Evolution. Great apes and zoonoses.
Paul M Sharp et al.
Science (New York, N.Y.), 340(6130), 284-286 (2013-04-20)
T A Timofeeva et al.
Voprosy virusologii, 58(1), 24-27 (2013-06-22)
The influenza virus hemagglutinin (HA) is an envelope virus glycoprotein responsible for the attachment of the virus particles to cells via binding terminal sialic acid residues of cell surface oligosaccharides. In our previous works on influenza A virus escape mutants
Sasirekha Ramani et al.
Journal of virology, 87(13), 7255-7264 (2013-04-26)
Naturally occurring bovine-human reassortant rotaviruses with a P[11] VP4 genotype exhibit a tropism for neonates. Interaction of the VP8* domain of the spike protein VP4 with sialic acid was thought to be the key mediator for rotavirus infectivity. However, recent
Influenza: Pathways to human adaptation.
David A Steinhauer
Nature, 499(7459), 412-413 (2013-07-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service