Skip to Content
Merck
All Photos(1)

Key Documents

A8648

Sigma-Aldrich

p-Aminophenyl β-D-thiogalactopyranoside–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106514

form

saline suspension

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

12 atoms

storage temp.

2-8°C

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service