Skip to Content
Merck
All Photos(1)

Key Documents

A7833

Sigma-Aldrich

Anti-ATF-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35-38 kDa

species reactivity

human

technique(s)

microarray: suitable
western blot (chemiluminescent): 1:500 using whole cell extract of human Jurkat acute T cell leukemia cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATF1(466)

General description

Activating Transcription Factor 1 (ATF1) or TREB-36 is a member of the ATF/ CREB (cAMP response element binding protein) family of basic region leucine-zipper (bZip) DNA-binding proteins. It is encoded by the gene mapped to human chromosome 12q13.12. ATF1 mRNA is expressed in variety of tissues.

Specificity

Anti-ATF1 specifically recognizes human ATF1.

Immunogen

synthetic peptide corresponding to human ATF-1, N-terminal sequence, amino acids 6-23, conjugated to KLH.

Application

Rabbit Anti-ATF-1 antibody is suitable for western blot applications at 1:500.

Biochem/physiol Actions

Activating transcription factor 1 (ATF1) mediates both Ca2+ and cAMP responses at several levels. It binds to the tax-responsive element (TRE1) of the human T-cell lymphotropic virus type-I (HTLV-1). ATF1 is detectable in metastatic melanoma cells and seems to contribute to their survival. A chimeric protein composed of the N-terminal domain of EWS (Ewing sarcoma oncogene) linked to the bZip domain of ATF1 is implicated in the rare malignant clear cell sarcoma of tendon sheath and aponeuroses (malignant melanoma of soft parts).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For prolonged storage, freeze in working aliquots at -20 °C. Repeated freezing and thawing is not recom-mended. Storage in “frost-free” freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activating transcription factor 1 and CREB are important for cell survival during early mouse development
Bleckmann SC, et al.
Molecular and Cellular Biology, 22(6), 1919-1925 (2002)
Systematic functional interrogation of genes in GWAS loci identified ATF1 as a key driver in colorectal cancer modulated by a promoter-enhancer interaction
Tian J, et al.
American Journal of Human Genetics, 105(1), 29-47 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service