Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

A2394

Sigma-Aldrich

Acetylcholine perchlorate

Synonym(s):

ACh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16ClNO6
CAS Number:
Molecular Weight:
245.66
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (TLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[O-]Cl(=O)(=O)=O.CC(=O)OCC[N+](C)(C)C

InChI

1S/C7H16NO2.ClHO4/c1-7(9)10-6-5-8(2,3)4;2-1(3,4)5/h5-6H2,1-4H3;(H,2,3,4,5)/q+1;/p-1

InChI key

SHIQLFRCVFUYEK-UHFFFAOYSA-M

Gene Information

human ... CHRM3(1131)

Application

Acetylcholine is an endogenous neurotransmitter at cholinergic synapses that amplifies action potential of the sarcolemma thereby inducing muscle contractions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

A2394-25G:
A2394-BULK:
A2394-5G:
A2394-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aswin Verhoeven et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 168(2), 314-326 (2004-05-14)
It is demonstrated that internuclear distances can be evaluated from rotational-resonance (RR) experiments in uniformly (13)C-labelled compounds. The errors in the obtained distances are less than 10% without the need to know any parameters of the spin system except the
Saikat Ray et al.
Science (New York, N.Y.), 343(6173), 891-896 (2014-01-25)
Little is known about how microcircuits are organized in layer 2 of the medial entorhinal cortex. We visualized principal cell microcircuits and determined cellular theta-rhythmicity in freely moving rats. Non-dentate-projecting, calbindin-positive pyramidal cells bundled dendrites together and formed patches arranged
Xun Qian et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(1), 127-135 (2013-11-02)
Intermediate and small conductance KCa channels IK1 (KCa3.1) and SK3 (KCa2.3) are primary targets of endothelial Ca(2+) signals in the arterial vasculature, and their ablation results in increased arterial tone and hypertension. Activation of IK1 channels by local Ca(2+) transients
Yingchuan B Qi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(12), 5319-5325 (2013-03-22)
Excitatory acetylcholine motor neurons drive Caenorhabditis elegans locomotion. Coordinating the activation states of the backward-driving A and forward-driving B class motor neurons is critical for generating sinusoidal and directional locomotion. Here, we show by in vivo calcium imaging that expression
Elim Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(52), 21171-21176 (2013-12-12)
The habenulo-interpeduncular pathway, a highly conserved cholinergic system, has emerged as a valuable model to study left-right asymmetry in the brain. In larval zebrafish, the bilaterally paired dorsal habenular nuclei (dHb) exhibit prominent left-right differences in their organization, gene expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service