Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

96151

Sigma-Aldrich

Nano beads based on PD, Chromeon 545-marked, carboxylated

Synonym(s):

PD-Chromeon 545, carboxylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

beads

fluorescence

λex 545 nm; λem 575 nm in H2O

storage temp.

2-8°C

Features and Benefits

PD is a new polymer, which provides less sensitivity to oxygen and reduced photo bleaching compared to latex (polystyrene) particles. PD particles are ideally suited for phosphorescent lifetime (decay time) measurements. Fluorescent PD particles are suitable for protein labeling and detection of antigen-antibody reactions.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

96151-10ML-F:
96151-BULK-F:
96151-5ML-F:
96151-VAR-F:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service