Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

89105

Sigma-Aldrich

4-Methylumbelliferyl-β-D-glucuronide

≥99.0% (HPLC)

Synonym(s):

MUGlcU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O9
CAS Number:
Molecular Weight:
352.29
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥99.0% (HPLC)

form

solid

impurities

≤0.1% free 4-methylumbelliferone
≤15% water

fluorescence

λem 445 nm±5 nm

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](O)[C@@H](C(O)=O)O[C@H]1OC2=CC(OC(C=C3C)=O)=C3C=C2

InChI

1S/C16H16O9/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22/h2-5,11-14,16,18-20H,1H3,(H,21,22)/t11-,12-,13+,14-,16+/m0/s1

InChI key

ARQXEQLMMNGFDU-JHZZJYKESA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as substrate for β-glucuronidase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

89105-BULK:
89105-25MG:
89105-25MG-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service