Skip to Content
Merck
All Photos(1)

Documents

Safety Information

78127

Sigma-Aldrich

2-Ethyl-1,6-dioxaspiro[4,4]nonane

≥96.0% (GC)

Synonym(s):

Chalcogran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥96.0% (GC)

form

liquid

refractive index

n20/D 1.446

SMILES string

CCC1CCC2(CCCO2)O1

InChI

1S/C9H16O2/c1-2-8-4-6-9(11-8)5-3-7-10-9/h8H,2-7H2,1H3

InChI key

DCWKALWZHORJAO-UHFFFAOYSA-N

Application

Park beetle pheromone, plant protectant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

78127-VAR:
78127-BULK:
78127-1G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Byers et al.
Die Naturwissenschaften, 87(11), 503-507 (2001-01-11)
Volatiles from leaves or bark of nonhost birch (Betula pendula) and Norway spruce (Picea abies) dramatically reduced the attraction of the bark beetle, Pityogenes bidentatus (Coleoptera: Scolytidae), to their aggregation pheromone components (cis-verbenol and grandisol) in the field. In addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service