Skip to Content
Merck
All Photos(1)

Documents

Safety Information

51667

Sigma-Aldrich

8-Heptadecene

≥96.0% (mixture of isomers (cis+trans), GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34
CAS Number:
Molecular Weight:
238.45
Beilstein:
1840926
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥96.0% (mixture of isomers (cis+trans), GC)

form

solid

SMILES string

CCCCCCCC\C=C\CCCCCCC

InChI

1S/C17H34/c1-3-5-7-9-11-13-15-17-16-14-12-10-8-6-4-2/h15,17H,3-14,16H2,1-2H3/b17-15+

InChI key

BIQKRILZMDQPHI-BMRADRMJSA-N

General description

may contain up to 15% trans-8-heptadecene and 1% heptadecane

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

51667-BULK:
51667-VAR:
51667-25MG:
51667-5MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid Maruti et al.
Journal of chromatography. A, 1551, 41-51 (2018-04-14)
A novel extraction technique is proposed in which the Multiple Headspace Extraction (MHE) approach is used in conjunction with Headspace Sorptive Extraction (HSSE) and Gas Chromatography-Mass Spectrometry (GC-MS) detection. The extraction method was developed to determine volatile compounds in macroalgae.
Marta Krasny et al.
Cell and tissue banking, 16(4), 631-638 (2015-07-15)
Increasingly dental surgeons face the challenge of reconstruction of the height and/or thickness of the alveolar ridge as more and more patients wish to have permanent restoration of their dental defects based on intraosseous implants. Evaluation of human allogeneic bone
Ki-Chang Lee et al.
Journal of agricultural and food chemistry, 68(9), 2803-2815 (2020-02-11)
This study evaluated the applicability of a rapid analytical method using a headspace solid-phase microextraction gas chromatography/mass spectrometry (HS-SPME-GC/MS) technique to identify gamma-irradiated soybeans (0.1-5 kGy). From the partial least squares discriminant analysis used to discriminate between non-irradiated and irradiated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service