312061
n-Butanol-1-14C
Synonym(s):
Butyl-1-14C alcohol
About This Item
extent of labeling
1-10 mCi per mmol
format
neat
storage temp.
2-8°C
SMILES string
CCC[14CH2]O
Looking for similar products? Visit Product Comparison Guide
Packaging
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service