Skip to Content
Merck
All Photos(1)

Documents

Safety Information

07602

Sigma-Aldrich

FSB staining solution

1 % (w/v) in DMSO, fluorescent stain for amyloid

Synonym(s):

1-Fluoro-2,5-bis[(E)-3-carboxy-4-hydroxystyryl]benzene solution, 3,3′-[(2-Fluoro-1,4-phenylene)di-(1E)-2,1-ethenediyl]bis[6-hydroxybenzoic acid] solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H17FO6
CAS Number:
Molecular Weight:
420.39
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

liquid

concentration

1 % (w/v) in DMSO

UV absorption

λ: 360-380 nm Amax: 0.6-0.9

storage temp.

2-8°C

SMILES string

OC(C1=CC(/C=C/C2=CC=C(/C=C/C3=CC(C(O)=O)=C(O)C=C3)C(F)=C2)=CC=C1O)=O

InChI

1S/C24H17FO6/c25-20-13-16(2-1-14-5-9-21(26)18(11-14)23(28)29)4-8-17(20)7-3-15-6-10-22(27)19(12-15)24(30)31/h1-13,26-27H,(H,28,29)(H,30,31)/b2-1+,7-3+

InChI key

DZXZHGCGNRRJEG-WMWQKROPSA-N

General description

1-Fluoro-2,5-bis[(E)-3-carboxy-4-hydroxystyryl]benzene solution (FSB ) is an amyloid-β peptide (Aβ) specific dye. It is a fluorine analog of 1-Bromo-2,5-bis(3-carboxy-4-hydroxystyryl)benzene (BSB). It also acts as an amyloidophilic fluorescent probe for Aβ.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water soluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

07602-0.1ML-BULK:
07602-0.1ML:
07602-BULK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FK506 reduces amyloid plaque burden and induces MMP-9 in AβPP/PS1 double transgenic mice.
Hong H S, et al.
Journal of Alzheimer'S Disease, 22(1), 97-105 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service