Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

27081

Sigma-Aldrich

Chromium(VI) oxide

puriss. p.a., ACS reagent, ≥99.0% (RT)

Synonym(s):

Chromic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
form:
powder, crystals or granules

grade

ACS reagent
puriss. p.a.

Assay

≥99.0% (RT)

form

powder, crystals or granules

impurities

≤0.005% total nitrogen (N)

pH

<1 (20 °C, 100 g/L)

mp

196 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤50 mg/kg
Ba: ≤50 mg/kg
Ca: ≤100 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cu: ≤10 mg/kg
Fe: ≤100 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤10 mg/kg
Na: ≤2000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Oxidant used in the decomposition by solubilization or fusion[1][2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Specified Class I Designated Chemical Substances

FSL

Group 1: Oxidizing solids
Oxidation products of chromium, lead, or iodine
Hazardous rank III
3rd oxidizing solid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

27081-VAR-F:
27081-50G-F:4548173219356
27081-BULK-F:
27081-1KG-F:4548173219332
27081-250G-F:4548173219349


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raman spectroscopy of chromium oxide supported on Al2O3, TiO2 and SiO2: A comparative study.
Hardcastle FD and Wachs IE.
J. Mol. Catal., 46(1), 173-186 (1988)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Study of the chromium (VI) oxide-pyridine complex.
Holum JR.
The Journal of Organic Chemistry, 26(12), 4814-4816 (1961)
Chromium (VI) oxide-catalyzed oxidation of arenes with periodic acid.
Yamazaki S.
Tetrahedron Letters, 42(19), 3355-3357 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service