Skip to Content
Merck
All Photos(1)

Documents

Safety Information

24553

Sigma-Aldrich

Petroleum ether

puriss., high boiling, bp 60-80 °C

Synonym(s):

Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502

vapor density

2.5 (vs air)

vapor pressure

25.8 psi

grade

puriss.

impurities

≤0.002% non-volatile matter
≤0.005% S-compounds (as S)
≤0.01% water (Karl Fischer)
≤0.02% benzene (GC)

refractive index

n20/D 1.376-1.382

bp

60-80 °C

density

0.660-0.680 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

General description

Petroleum ether is a high boiling point fraction having a boiling point in the range of 60-80°C. Extracts of leaves of Litchi chinensis Gaertn. (Sapindaceae) have been prepared in petroleum ether and were evaluated for antiinflammatory, analgesic and antipyretic activities.

Application

Petroleum ether may be employed in the extraction of biologically active carotenoid pigments from the total carotenoid pigments by adsorption. It may be employed in the solvent reservoir during the separation of lipid mixtures by adsorption chromatography with silicic acid.

Other Notes

Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-4.0 °F

Flash Point(C)

< -20 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

24553-25L-R:
24553-5L-R:4548173962153
24553-BULK-R:
24553-4X5L-R:
24553-VAR-R:
24553-1L-R:4548173962146
24553-6X1L-R:
24553-2.5L-R:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranganna S.
Handbook of Analysis and Quality Control for Fruit and Vegetable Products, 84-84 (1986)
The separation of complex lipide mixtures by the use of silicic acid chromatography.
J HIRSCH et al.
The Journal of biological chemistry, 233(2), 311-20 20-311-20 20 (1958-08-01)
S E Besra et al.
Journal of ethnopharmacology, 54(1), 1-6 (1996-10-01)
Pharmacological studies were conducted with the petroleum ether extract of leaves of the plant Litchi chinensis Gaertn. (Sapindaceae) on experimental animals. The extract was found to possess antiinflammatory, analgesic and antipyretic activity. Acute toxicity studies revealed that the extract, up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service