Skip to Content
Merck
All Photos(1)

Documents

Safety Information

Y0002024

Plastic additive 27

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dioctyl terephthalate, Bis(2-ethylhexyl) terephthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-[CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
390.56
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

1 mmHg ( 217 °C)

manufacturer/tradename

EDQM

refractive index

n20/D 1.49 (lit.)

bp

400 °C (lit.)

mp

−48 °C

density

0.986 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

CCCCC(CC)COC(=O)c1ccc(cc1)C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-11-19(7-3)17-27-23(25)21-13-15-22(16-14-21)24(26)28-18-20(8-4)12-10-6-2/h13-16,19-20H,5-12,17-18H2,1-4H3

InChI key

RWPICVVBGZBXNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Plastic additive 27 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

413.6 °F - closed cup

Flash Point(C)

212 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 4 petroleums
Hazardous rank III

JAN Code

Y0002024:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service