Skip to Content
Merck
All Photos(1)

Documents

Safety Information

RP-SA1-1

Recombinant Sin a 1 (Sinapis alba allergen 1.0101), Mustard

pkg of 250 μg, storage temp.:-20°C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.26

grade

analytical standard

Assay

>95%

mol wt

17 kDa

packaging

pkg of 250 μg

manufacturer/tradename

InBio

technique(s)

HPLC: suitable
LC/MS: suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

General description

Produced and qualified by Indoor Biotechnologies.

Source: Pichia pastoris
Formulation: Preservative and carrier-free in 1X PBS, 0.3M NaCl, pH 7.4. Filtered through 0.22μm filter.
Concentration: See product insert.
Purification: Purified from Pichia pastoris culture by multi-step column chromatography. Purity > 95% by silver stained SDS-PAGE.

Note:
(1) rSin a 1 has a C-terminal 6xHis-tag.
(2) Avoid repeated Freeze/Thaw cycles. It is recommended to aliquot the product upon first thaw and freeze in working aliquot volumes.

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

RP-SA1-1:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service