Recommended Products
grade
certified reference material
Quality Level
manufacturer/tradename
RTC MDL-003
concentration
in H2O
technique(s)
TOC: suitable
application(s)
environmental
format
matrix material
General description
To prepare solution, partially fill a 1-L volumetric flask with reagent-grade water.Open ampule MDL-003 (be careful to avoid cuts on glass) and transfer 10 mL from the ampule into the flask. Fill the volumetric flask to volume with reagent-grade water and mix well. The sample is now ready for immediate analysis.
NOTES: For proper oxygen depletion of BOD, the sample should be tested in a dilution series. Use a natural surface water or diluted domestic sewage for seeded dilution water and avoid entrainment of air. Be sure to etermine the “blank” BOD of your seeded dilution water so that the proper seed correction can be made.
Other Notes
Analyte
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
MDL003-100ML:
MDL003-20ML:
MDL003-20ML-PW:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service