Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

M4625

Sigma-Aldrich

D-(+)-Mannose

Synonym(s):

D-Mannopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1564373
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

99% (GC)

mp

133-140 °C (lit.)

SMILES string

OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1

InChI key

WQZGKKKJIJFFOK-QTVWNMPRSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

M4625-VAR:
M4625-100G:
M4625-5G:
M4625-5G-KC:
M4625-500G:
M4625-25G:
M4625-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Otter et al.
Hepatology (Baltimore, Md.), 16(1), 54-59 (1992-07-01)
Various studies have shown that mannose receptors rapidly eliminate glycoproteins and microorganisms bearing high mannose-type carbohydrate chains from the blood circulation. The purpose of this study was to characterize the mannose receptor in the liver, which in vivo is involved
Chia-Suei Hung et al.
Molecular microbiology, 44(4), 903-915 (2002-05-16)
The first step in the colonization of the human urinary tract by pathogenic Escherichia coli is the mannose-sensitive binding of FimH, the adhesin present at the tip of type 1 pili, to the bladder epithelium. We elucidated crystallographically the interactions
A T Van der Ploeg et al.
The Journal of clinical investigation, 87(2), 513-518 (1991-02-01)
The lysosomal storage disorder glycogenosis type II is caused by acid alpha-glucosidase deficiency. In this study we have investigated the possible applicability of mannose 6-phosphate receptor-mediated enzyme replacement therapy to correct the enzyme deficiency in the most affected tissues. Bovine
M R Bürk et al.
Placenta, 22(4), 309-316 (2001-04-05)
Pre-eclampsia is a disorder of unknown aetiology peculiar to human pregnancy. A well-described pathological feature being shallow trophoblast invasion into the spiral arteries during placenta development. Epidemiological studies have revealed an increased risk in pregnancies of primipaternity, and an association
I Adlerberth et al.
Applied and environmental microbiology, 62(7), 2244-2251 (1996-07-01)
Two Lactobacillus plantarum strains of human intestinal origin, strains 299 (= DSM 6595) and 299v (= DSM 9843), have proved to be efficient colonizers of the human intestine under experimental conditions. These strains and 17 other L. plantarum strains were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service