LTN-AH3-1
LoTox Natural Ara h 3 (Arachis hypogaea allergen 3), peanut
pkg of 250 μg, storage temp.:-20°C
About This Item
Recommended Products
grade
analytical standard
Assay
>95%
mol wt
(Multiple subunits ranging from 14-45 kDa)
packaging
pkg of 250 μg
manufacturer/tradename
InBio
technique(s)
HPLC: suitable
LC/MS: suitable
application(s)
food and beverages
format
neat
storage temp.
−20°C
General description
Source: Virginia raw peanuts
Formulation: Preservative-free and carrier-free in 20mM Tris, pH 8.2, 0.15M NaCl. Sterile filtered
Concentration: See product insert
Purification: Purified from peanut extract by multi- step chromatography. Purity > 95 % by silver stained SDS-PAGE, reducing conditions.
Note:
(1) A LoTox product, Endotoxin < 0.03 EU/μg.
(2) Avoid repeated Freeze/Thaw cycles. It is recommended to aliquot the product upon first thaw and freeze in working aliquot volumes.
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
LTN-AH3-1:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service