Skip to Content
Merck
All Photos(1)

Documents

Safety Information

ERMBC190

Rapeseed (S, total glucosinolate)

ERM®, certified reference material, medium level

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

ERM®
certified by the BSC

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Packaging

package with 20 g

Analysis Note

For more information please see:
ERMBC190

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Total glucosinolate (GSL)
Trace Elements: Sulphur

Matrix Group: Nuts, Soy, Edible Oils and Fats

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

ERMBC190-20G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service