BCR644
Artificial foodstuff (Fructose, Sucrose, Lactose and Starch/glucose)
BCR®, certified reference material
About This Item
Recommended Products
grade
certified reference material
Agency
BCR®
form
solid
manufacturer/tradename
JRC
application(s)
food and beverages
format
matrix material
storage temp.
2-8°C
Analysis Note
BCR644
Other Notes
Carbohydrates: Fructose, Lactose, Starch/glucose, Sucrose
Matrix Group: Processed Food
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
BCR644-50G:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service