Skip to Content
Merck
All Photos(1)

Key Documents

92070

Supelco

Suberoyl-L-carnitine

≥97.0% (TLC)

Synonym(s):

(2R)-3-Carboxy-2-[(7-carboxy-1-oxoheptyl)oxy]-N,N,N-trimethyl-1-propanaminium inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H27NO6
CAS Number:
Molecular Weight:
317.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥97.0% (TLC)

optical activity

[α]/D -15±2°, c = 0.1 in H2O

impurities

≤10% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C\C=C/C=C
C[N+](C)(C)C[C@H](OC(CCCCCCC(O)=O)=O)CC([O-])=O

InChI

1S/C15H27NO6/c1-16(2,3)11-12(10-14(19)20)22-15(21)9-7-5-4-6-8-13(17)18/h12H,4-11H2,1-3H3,(H-,17,18,19,20)/t12-/m1/s1

InChI key

YVWVEIPYMGBQPE-GFCCVEGCSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service