Skip to Content
Merck
All Photos(1)

Key Documents

91651

Sigma-Aldrich

Triethyloxonium hexafluorophosphate

purum, ≥95.0% (NMR)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3OPF6
CAS Number:
Molecular Weight:
248.15
Beilstein:
4302183
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

purum

Assay

≥95.0% (NMR)

mp

~145 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

CC[O+](CC)CC.F[P-](F)(F)(F)(F)F

InChI

1S/C6H15O.F6P/c1-4-7(5-2)6-3;1-7(2,3,4,5)6/h4-6H2,1-3H3;/q+1;-1

InChI key

JRHIFBONIQGZFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent involved in:
  • Synthesis of ruthenium allenylidene complexes
  • Asymmetric Diels-Alder reactions
  • Chlorine scavenging
  • Synthesis of molecules for studies of wavelength tunability via cis-trans isomerization
  • Reactions with 2-mthylpyridine N-oxide

Catalyst activating agent for asymmetric imine aziridination via diazo ester activation

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service