915971
Dirhodium(II)tetrakisoctanoate
Synonym(s):
Rhodium(II) octanoate, dimer, [Rh(C7H15COO)2]2
About This Item
Recommended Products
form
powder
Quality Level
mp
289-292 °C
SMILES string
CCCCCCCC(=O)O[Rh](OC(=O)CCCCCCC)[Rh](OC(=O)CCCCCCC)OC(=O)CCCCCCC
InChI
1S/4C8H16O2.2Rh/c4*1-2-3-4-5-6-7-8(9)10;;/h4*2-7H2,1H3,(H,9,10);;/q;;;;2*+2/p-4
InChI key
FZXFNYFVNKTQSX-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Storage and Stability
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
915971-BULK:
915971-VAR:
915971-1G:4548173372846
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service