Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

87638

Sigma-Aldrich

5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin iron(III) chloride complex

purum, ≥98.0% (HPLC)

Synonym(s):

meso -Tetrakis(pentafluorophenyl)porphyrin iron(III) chloride, Chloro[5,10,15,20-tetrakis(pentafluorophenyl)-21H ,23H-porphinato]iron(III)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H8ClF20FeN4
CAS Number:
Molecular Weight:
1063.83
Beilstein:
8674334
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

purum

Assay

≥98.0% (HPLC)

SMILES string

Fc1c(F)c(F)c(c(F)c1F)-c2c3ccc(n3)c(-c4c(F)c(F)c(F)c(F)c4F)c5ccc6c(-c7c(F)c(F)c(F)c(F)c7F)c8ccc(n8)c(-c9c(F)c(F)c(F)c(F)c9F)c%10ccc2n%10[Fe](Cl)n56

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Electron-deficient iron-porphyrin complex which is more stable to oxidation than iron-TPP; Employed as a cytochrome-model in oxidation reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

87638-100MG:
87638-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.J. Castellino et al.
Journal of the American Chemical Society, 110, 158-158 (1988)
Y. Watanabe et al.
Journal of the American Chemical Society, 111, 8047-8047 (1989)
Journal of the American Chemical Society, 110, 7512-7512 (1988)
C.K. Chang et al.
Journal of the Chemical Society. Chemical Communications, 778-778 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service