Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

86628

Supelco

Tetraethylammonium hydrogen sulfate

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(HSO4)
CAS Number:
Molecular Weight:
227.32
Beilstein:
6590581
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

Assay

≥98.0% (T)

impurities

≤2% water

mp

240-245 °C

SMILES string

OS([O-])(=O)=O.CC[N+](CC)(CC)CC

InChI

1S/C8H20N.H2O4S/c1-5-9(6-2,7-3)8-4;1-5(2,3)4/h5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

CREVBWLEPKAZBH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

86628-50G:
86628-BULK:
86628-250G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service