Skip to Content
Merck
All Photos(1)

Key Documents

80634

Sigma-Aldrich

PIPES dipotassium salt

≥99.0% (T)

Synonym(s):

1,4-Piperazinediethanesulfonic acid dipotassium salt, Piperazine-1,4-bis(2-ethanesulfonic acid) dipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16K2N2O6S2
CAS Number:
Molecular Weight:
378.55
MDL number:
UNSPSC Code:
12161700

Assay

≥99.0% (T)

impurities

≤10% water

useful pH range

6.1-7.5

pKa (25 °C)

6.8

solubility

H2O: 0.5 g/mL, clear, colorless

SMILES string

[K+].[K+].[O-]S(=O)(=O)CCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O6S2.2K/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;;/h1-8H2,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2

InChI key

BJQYFCAMUXGZFN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service