Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

75833

Supelco

Prenyl acetate

analytical standard

Synonym(s):

3,3-dimethyl allyl acetate, 3-methyl-2-buten-1-ol acetate, 3-methyl-2-buten-1-yl acetate, 3-methyl-2-butenyl acetate, Isopent-2-enyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.428-1.432
n20/D 1.43 (lit.)

bp

151-152 °C/752 mmHg (lit.)

density

0.917 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)OC\C=C(\C)C

InChI

1S/C7H12O2/c1-6(2)4-5-9-7(3)8/h4H,5H2,1-3H3

InChI key

XXIKYCPRDXIMQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

75833-VAR:
75833-1ML:
75833-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric M Jones et al.
Cell systems, 8(3), 254-260 (2019-03-25)
G protein-coupled receptors (GPCRs) are central to how mammalian cells sense and respond to chemicals. Mammalian olfactory receptors (ORs), the largest family of GPCRs, mediate the sense of smell through activation by small molecules, though for most bonafide ligands, they

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service